Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Anisaldehyde, 99+%

Catalog No. AC104800025
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104800025 2.5 L Glass bottle
AC104800050 5 mL Glass bottle
AC104801000 100 mL Glass bottle
AC104805000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC104800025 Supplier Thermo Scientific Chemicals Supplier No. 104800025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 123-11-5 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-11-5
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00003385
InChI Key ZRSNZINYAWTAHE-UHFFFAOYSA-N
Synonym p-anisaldehyde, anisaldehyde, anisic aldehyde, p-methoxybenzaldehyde, 4-anisaldehyde, aubepine, p-anisic aldehyde, p-formylanisole, benzaldehyde, 4-methoxy, crategine
PubChem CID 31244
ChEBI CHEBI:28235
IUPAC Name 4-methoxybenzaldehyde
SMILES COC1=CC=C(C=C1)C=O

Specifications

Melting Point -1°C
Color Colorless to Yellow
Density 1.1210g/mL
Boiling Point 248°C to 249°C
Flash Point 116°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5710 to 1.574
Linear Formula CH3OC6H4CHO
Quantity 2.5 L
Beilstein 08,67
Specific Gravity 1.121
Merck Index 15,656
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: miscible withe alcohol and ether,soluble in 5 volumes of 50% alcohol,soluble in most organic solvents
Viscosity 4.1 mPa.s (25°C)
Formula Weight 136.15
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material p-Anisaldehyde, 99+%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-602-6
RTECSNumber BZ2625000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.