Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Anisaldehyde, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012818 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2818 5 mL
11-101-2817 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2818 Supplier MilliporeSigma Supelco Supplier No. 970635MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

p-anisaldehyde is an aromatic aldehyde commonly found in anise seed oil. It shows acaricidal activity and is primarily used, as a lead compound for the development of new agents for the selective control of house dust mites.

TRUSTED_SUSTAINABILITY

Specifications

CAS 123-11-5
Vapor Density 4.7 (vs. air)
Density 1.119 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3OC6H4CHO
MDL Number MFCD00003385
Melting Point -1°C
Boiling Point 248°C
Synonym 4-Methoxybenzaldehyde; Aubépine
Quantity 5 mL
Formula Weight 136.15
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.