Learn More
Medchemexpress LLC p-Anisic acid | 100-09-4 | 152.15 | 10 G
SDP
Catalog No. 5000415802
Encompass_Preferred
p-Anisic acid (4-Methoxybenzoic acid) is an orally available tyrosinase inhibitor with antioxidant, anti-anxiety, anti-inflammatory, anti-tumor, anti-diabetic, and preservative properties. It can also be used as a preservative in cosmetics.
  • Orally available tyrosinase inhibitor
  • Exhibits antioxidant properties
  • Has anti-anxiety effects
  • Possesses anti-inflammatory properties
  • Shows anti-tumor activity
  • Has anti-diabetic effects, including blood sugar-lowering and improvement of pancreatic morphology
  • Acts as a preservative, useful in the cosmetics field
  • Initial source: plants (leguminosae acacia mearnsii)
  • Endogenous metabolite
Catalog No. 50-004-15802 Supplier Medchemexpress LLC Supplier No. HYN139410G
p-Anisic acid (4-Methoxybenzoic acid) is an orally available tyrosinase inhibitor with antioxidant, anti-anxiety, anti-inflammatory, anti-tumor, anti-diabetic, and preservative properties. It can also be used as a preservative in cosmetics.
  • Orally available tyrosinase inhibitor
  • Exhibits antioxidant properties
  • Has anti-anxiety effects
  • Possesses anti-inflammatory properties
  • Shows anti-tumor activity
  • Has anti-diabetic effects, including blood sugar-lowering and improvement of pancreatic morphology
  • Acts as a preservative, useful in the cosmetics field
  • Initial source: plants (leguminosae acacia mearnsii)
  • Endogenous metabolite