Learn More
Medchemexpress LLC p-Anisic acid | 100-09-4 | 152.15 | 100 G
SDP
Catalog No. 5000415113
Encompass_Preferred
p-Anisic acid is an orally available tyrosinase inhibitor with antioxidant, anti-anxiety, anti-inflammatory, anti-tumor, anti-diabetic, and preservative properties. It can be utilized as a preservative in the cosmetics field.
  • Orally available
  • Tyrosinase inhibitor
  • Antioxidant properties
  • Anti-anxiety properties
  • Anti-inflammatory properties
  • Anti-tumor properties
  • Anti-diabetic properties
  • Preservative properties
Catalog No. 50-004-15113 Supplier Medchemexpress LLC Supplier No. HYN1394100G
p-Anisic acid is an orally available tyrosinase inhibitor with antioxidant, anti-anxiety, anti-inflammatory, anti-tumor, anti-diabetic, and preservative properties. It can be utilized as a preservative in the cosmetics field.
  • Orally available
  • Tyrosinase inhibitor
  • Antioxidant properties
  • Anti-anxiety properties
  • Anti-inflammatory properties
  • Anti-tumor properties
  • Anti-diabetic properties
  • Preservative properties