Learn More
Medchemexpress LLC p-Anisic acid (Standard) | 100-09-4 | 152.15 | 100 MG
SDP
Catalog No. 5000416147
Encompass_Preferred
p-Anisic acid (Standard) is an analytical standard used in research and analytical applications. This compound, also known as 4-Methoxybenzoic acid, possesses anti-bacterial and antiseptic properties. It serves as a reference standard for various experimental methodologies.
  • Analytical standard for p-Anisic acid
  • Intended for research and analytical applications
  • Possesses anti-bacterial and antiseptic properties
  • Used in qualitative, quantitative, and methodological research experiments
  • Commonly applied in HPLC, GC, and MS studies
Catalog No. 50-004-16147 Supplier Medchemexpress LLC Supplier No. HYN1394R100MG
p-Anisic acid (Standard) is an analytical standard used in research and analytical applications. This compound, also known as 4-Methoxybenzoic acid, possesses anti-bacterial and antiseptic properties. It serves as a reference standard for various experimental methodologies.
  • Analytical standard for p-Anisic acid
  • Intended for research and analytical applications
  • Possesses anti-bacterial and antiseptic properties
  • Used in qualitative, quantitative, and methodological research experiments
  • Commonly applied in HPLC, GC, and MS studies