Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Anisoyl chloride, 99%

Catalog No. AC104855000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC104855000 500 g
AC104850050 5 g
AC104851000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC104855000 Supplier Thermo Scientific Chemicals Supplier No. 104855000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 100-07-2 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-07-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00000687
InChI Key MXMOTZIXVICDSD-UHFFFAOYSA-N
Synonym p-anisoyl chloride, benzoyl chloride, 4-methoxy, anisoyl chloride, 4-anisoyl chloride, p-methoxybenzoyl chloride, 4-methoxybenzoylchloride, 4-methoxybenzoic acid chloride, benzoyl chloride, p-methoxy, p-methoxybenzoic acid chloride, 4-methoxy-benzoyl chloride
PubChem CID 7477
IUPAC Name 4-methoxybenzoyl chloride
SMILES COC1=CC=C(C=C1)C(Cl)=O

Specifications

Color Colorless to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5800 to 1.5820 (20°C, 589nm)
Linear Formula CH3OC6H4COCl
Quantity 500 g
Beilstein 10, 163
Merck Index 15, 665
Formula Weight 170.6
Percent Purity ≥98.5%
Physical Form Liquid After Melting
Chemical Name or Material p-Anisoyl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-816-4
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.