Learn More
Cayman Chemical p-APMSF (hydrochloride), Purity: ≥95%, CAS: 74938-88-8, 10 mg
SDP
Catalog No. 502970703
Encompass_Preferred
An irreversible inhibitor of serine proteases (Kis = 1.02, 1.18, 1.5, and 1.54 µM for bovine trypsin, human thrombin, bovine plasmin, and bovine Factor Xa, respectively); selective for these proteases over bovine chymotrypsin and acetylcholinesterase
Catalog No. 50-297-0703 Supplier Cayman Chemical Supplier No. 1497110 MG
An irreversible inhibitor of serine proteases (Kis = 1.02, 1.18, 1.5, and 1.54 µM for bovine trypsin, human thrombin, bovine plasmin, and bovine Factor Xa, respectively); selective for these proteases over bovine chymotrypsin and acetylcholinesterase