Learn More
Cayman Chemical p-Benzoyl-L-phenylalanine, Purity: ≥95%, CAS: 104504-45-2, 1 g
SDP
Catalog No. 502911952
Encompass_Preferred
A photoreactive derivative of L-phenylalanine; can be incorporated into synthetic peptides and used as a photolabel to identify the residues involved in peptide-protein interactions; has been incorporated into GPCRs to identify potential ligand binding sites
Catalog No. 50-291-1952 Supplier Cayman Chemical Supplier No. 361681 G
A photoreactive derivative of L-phenylalanine; can be incorporated into synthetic peptides and used as a photolabel to identify the residues involved in peptide-protein interactions; has been incorporated into GPCRs to identify potential ligand binding sites