Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Bromobenzoic Acid, MP Biomedicals

Catalog No. ICN205324 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205324 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205324 Supplier MP Biomedicals, Inc Supplier No. 0520532410

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 586-76-5
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.02
MDL Number MFCD00002529
InChI Key TUXYZHVUPGXXQG-UHFFFAOYSA-N
Synonym p-bromobenzoic acid, benzoic acid, 4-bromo, p-carboxybromobenzene, benzoic acid, p-bromo, p-bromobenzenecarboxylic acid, 4-bromo-benzoic acid, 4-brom-benzoesaeure, 4-carboxybromobenzene, para bromo benzoic acid, 4bromo-benzoic acid
PubChem CID 11464
ChEBI CHEBI:60698
IUPAC Name 4-bromobenzoic acid
SMILES OC(=O)C1=CC=C(Br)C=C1

Specifications

Quantity 10 g
Formula Weight 201.02
Chemical Name or Material p-Bromobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.