Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-P-Bromolevamisole Oxalate

Catalog No. 19004725MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-004-725MG 25 mg
19-004-7100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-004-725MG Supplier Sigma Aldrich Supplier No. 19004725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(-)-P-Bromolevamisole Oxalate

Specifications

CAS 62284-79-1
Melting Point 192°C
Molecular Formula C11H11BrN2S -+ C2H2O4
Linear Formula C11H11BrN2S -+ C2H2O4
MDL Number MFCD00070406
Quantity 25 mg
Synonym (-)-4-Bromotetramisole oxalate; 6-Bromolevamisole oxalate; S(-)-6-(4-Bromophenyl)-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole oxalate
Molecular Weight (g/mol) 373.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.