Learn More
Alkali Scientific ( ) p Bromotetramisole Oxalate, 25 Mg
SDP
Catalog No. 5000660000
Encompass_Preferred
Bromotetramisole Oxalate (25 mg) is a potent enzyme inhibitor used primarily as a phosphatase inhibitor in biochemical research. It is commonly used in studies focused on the regulation of phosphorylation events within cells. By inhibiting specific phosphatases, this reagent helps to preserve the phosphorylation state of proteins, facilitating research into signal transduction, enzyme activity, and cellular regulation. Bromotetramisole Oxalate is ideal for applications in proteomics, enzymology, and cellular biology, allowing for the detailed investigation of phosphatase function and its role in various cellular processes.
Catalog No. 50-006-60000 Supplier Alkali Scientific Supplier No. PBR002CP
Bromotetramisole Oxalate (25 mg) is a potent enzyme inhibitor used primarily as a phosphatase inhibitor in biochemical research. It is commonly used in studies focused on the regulation of phosphorylation events within cells. By inhibiting specific phosphatases, this reagent helps to preserve the phosphorylation state of proteins, facilitating research into signal transduction, enzyme activity, and cellular regulation. Bromotetramisole Oxalate is ideal for applications in proteomics, enzymology, and cellular biology, allowing for the detailed investigation of phosphatase function and its role in various cellular processes.