Learn More
Enzo Life Sciences (-)-p-Bromotetramisole. oxalate (5mg). CAS: 62284-79-1
SDP
Catalog No. 502009156
Encompass_Preferred
Potent alkaline phosphatase inhibitor. Mimics the action of orthovanadate in the potentiation of fluorouracil antiproliferative activity. Cell permeable.  Alternative name: (-)-(S)-6-(4-Bromophenyl)-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole . oxalate, L-p-Bromotetramisole . oxalate. Purity: ≥98% (HPLC). Solubility: Soluble in water, methanol, DMF or DMSO. Long Term Storage: +4°C.
Catalog No. 50-200-9156 Supplier Enzo Life Sciences Supplier No. ALX430025M005
Potent alkaline phosphatase inhibitor. Mimics the action of orthovanadate in the potentiation of fluorouracil antiproliferative activity. Cell permeable.  Alternative name: (-)-(S)-6-(4-Bromophenyl)-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole . oxalate, L-p-Bromotetramisole . oxalate. Purity: ≥98% (HPLC). Solubility: Soluble in water, methanol, DMF or DMSO. Long Term Storage: +4°C.