Learn More
Medchemexpress LLC (-)-p-Bromotetramisole oxalate | 62284-79-1 | 99.8% | 373.22 | 5 MG
SDP
Catalog No. 5000403349
Encompass_Preferred
(-)-p-Bromotetramisole oxalate is an alkaline phosphatase inhibitor. It is applicable to research related to erythroleukemia. In vitro, (-)-p-Bromotetramisole oxalate (0.3-0.4 mM; 5 days) potently inhibits terminal cell division-associated MEL DS-19 cell differentiation induced by 0.7% DMSO or 1.2 mM HMBA, reducing B+ cell levels. It also potently inhibits purified human liver, bone, spleen, and kidney alkaline phosphatase isoenzymes (10^-5-10^-4 mol/liter; under assay conditions at 30 °C) while minimally inhibiting purified human intestinal and placental alkaline phosphatase isoenzymes.
  • Alkaline phosphatase inhibitor.
  • Applicable to research related to erythroleukemia.
  • Potently inhibits terminal cell division-associated MEL DS-19 cell differentiation.
  • Potently inhibits purified human liver, bone, spleen, and kidney alkaline phosphatase isoenzymes.
Catalog No. 50-004-03349 Supplier Medchemexpress LLC Supplier No. HY196955MG
(-)-p-Bromotetramisole oxalate is an alkaline phosphatase inhibitor. It is applicable to research related to erythroleukemia. In vitro, (-)-p-Bromotetramisole oxalate (0.3-0.4 mM; 5 days) potently inhibits terminal cell division-associated MEL DS-19 cell differentiation induced by 0.7% DMSO or 1.2 mM HMBA, reducing B+ cell levels. It also potently inhibits purified human liver, bone, spleen, and kidney alkaline phosphatase isoenzymes (10^-5-10^-4 mol/liter; under assay conditions at 30 °C) while minimally inhibiting purified human intestinal and placental alkaline phosphatase isoenzymes.
  • Alkaline phosphatase inhibitor.
  • Applicable to research related to erythroleukemia.
  • Potently inhibits terminal cell division-associated MEL DS-19 cell differentiation.
  • Potently inhibits purified human liver, bone, spleen, and kidney alkaline phosphatase isoenzymes.