Learn More
Medchemexpress LLC (-)-p-Bromotetramisole oxalate | 62284-79-1 | 99.9% | 373.22 | 25 MG
SDP
Catalog No. 5000402413
Encompass_Preferred
(-)-p-Bromotetramisole oxalate (L-p-Bromotetramisole oxalate; 6-Bromolevamisole oxalate) is an alkaline phosphatase inhibitor. It is applicable for research related to erythroleukemia. It potently inhibits purified human liver, bone, spleen, and kidney alkaline phosphatase isoenzymes.
  • Inhibits terminal cell division-associated MEL DS-19 cell differentiation
  • Reduces B+ cell levels
  • Minimally inhibits purified human intestinal and placental alkaline phosphatase isoenzymes
Catalog No. 50-004-02413 Supplier Medchemexpress LLC Supplier No. 25MGHY19695
(-)-p-Bromotetramisole oxalate (L-p-Bromotetramisole oxalate; 6-Bromolevamisole oxalate) is an alkaline phosphatase inhibitor. It is applicable for research related to erythroleukemia. It potently inhibits purified human liver, bone, spleen, and kidney alkaline phosphatase isoenzymes.
  • Inhibits terminal cell division-associated MEL DS-19 cell differentiation
  • Reduces B+ cell levels
  • Minimally inhibits purified human intestinal and placental alkaline phosphatase isoenzymes