Learn More
Research Products International Corp RPI (-)-p-Bromotetramisole Oxalate, 25 Milligrams
SDP
Catalog No. 501977385
Encompass_Preferred
Potent alkaline phosphate inhibitor. Mimics the action of orthovanadate in the potentiation of fluorouracil antiproliferative activity. Cell permeable.
Catalog No. 50-197-7385 Supplier Research Products International Corp Supplier No. B743000.025
Potent alkaline phosphate inhibitor. Mimics the action of orthovanadate in the potentiation of fluorouracil antiproliferative activity. Cell permeable.