Learn More
Medchemexpress LLC P-CAB agent 2 | 1978371-23-1 | 98.0% | 432.51 g/mol | C22H25FN2O4S | 10 MG
SDP
Catalog No. 5000287075
Encompass_Preferred
P-CAB agent 2 is a potent, orally active potassium-competitive acid blocker provided as a research reagent for preclinical studies. It inhibits gastric H+/K+-ATPase with sub-100 nM potency and has reported activity in models of histamine-induced gastric acid secretion.
  • Potent H+/K+-ATPase inhibitor (IC50 <100 nM).
  • Orally active in preclinical models and inhibits histamine-induced gastric acid secretion.
  • Documented 98.0% purity suitable for research applications.
  • Analytical characterization available: COA, HNMR, LCMS, and SDS.
  • Provided in milligram-scale quantities for in vitro and in vivo studies.
  • Soluble in DMSO for assay preparation.
Catalog No. 50-002-87075 Supplier Medchemexpress LLC Supplier No. HY153219A10MG
P-CAB agent 2 is a potent, orally active potassium-competitive acid blocker provided as a research reagent for preclinical studies. It inhibits gastric H+/K+-ATPase with sub-100 nM potency and has reported activity in models of histamine-induced gastric acid secretion.
  • Potent H+/K+-ATPase inhibitor (IC50 <100 nM).
  • Orally active in preclinical models and inhibits histamine-induced gastric acid secretion.
  • Documented 98.0% purity suitable for research applications.
  • Analytical characterization available: COA, HNMR, LCMS, and SDS.
  • Provided in milligram-scale quantities for in vitro and in vivo studies.
  • Soluble in DMSO for assay preparation.