Learn More
Medchemexpress LLC P-CAB agent 2 | 1978371-23-1 | C22H25FN2O4S | 100 MG
SDP
Catalog No. 5000373987
Encompass_Preferred
P-CAB agent 2 is a potent and orally active potassium-competitive acid blocker and a gastric acid secretion inhibitor. It inhibits H+/K+-ATPase activity with an IC50 value of <100 nM and the hERG potassium channel with an IC50 value of 18.69 M. This agent shows no acute toxicity and inhibits histamine-induced gastric acid secretion.
  • Potent and orally active potassium-competitive acid blocker
  • Inhibits H+/K+-ATPase activity
  • Inhibits hERG potassium channel
  • Shows no acute toxicity
  • Inhibits histamine-induced gastric acid secretion
Catalog No. 50-003-73987 Supplier Medchemexpress LLC Supplier No. HY153219A100MG
P-CAB agent 2 is a potent and orally active potassium-competitive acid blocker and a gastric acid secretion inhibitor. It inhibits H+/K+-ATPase activity with an IC50 value of <100 nM and the hERG potassium channel with an IC50 value of 18.69 M. This agent shows no acute toxicity and inhibits histamine-induced gastric acid secretion.
  • Potent and orally active potassium-competitive acid blocker
  • Inhibits H+/K+-ATPase activity
  • Inhibits hERG potassium channel
  • Shows no acute toxicity
  • Inhibits histamine-induced gastric acid secretion