Learn More
Medchemexpress LLC P-CAB agent 2 hydrochloride | 2209911-80-6 | C22H26ClFN2O4S | 100 MG
SDP
Catalog No. 5000372539
Encompass_Preferred
P-CAB agent 2 hydrochloride is a potent and orally active potassium-competitive acid blocker and a gastric acid secretion inhibitor. It inhibits H+/K+-ATPase activity with an IC50 value of <100 nM and also inhibits the hERG potassium channel with an IC50 value of 18.69 μM. It shows no acute toxicity and inhibits histamine-induced gastric acid secretion.
  • Potent and orally active potassium-competitive acid blocker
  • Gastric acid secretion inhibitor
  • Inhibits H+/K+-ATPase activity
  • Inhibits hERG potassium channel
  • Shows no acute toxicity
  • Inhibits histamine-induced gastric acid secretion
  • For research use only
Catalog No. 50-003-72539 Supplier Medchemexpress LLC Supplier No. HY153219100MG
P-CAB agent 2 hydrochloride is a potent and orally active potassium-competitive acid blocker and a gastric acid secretion inhibitor. It inhibits H+/K+-ATPase activity with an IC50 value of <100 nM and also inhibits the hERG potassium channel with an IC50 value of 18.69 μM. It shows no acute toxicity and inhibits histamine-induced gastric acid secretion.
  • Potent and orally active potassium-competitive acid blocker
  • Gastric acid secretion inhibitor
  • Inhibits H+/K+-ATPase activity
  • Inhibits hERG potassium channel
  • Shows no acute toxicity
  • Inhibits histamine-induced gastric acid secretion
  • For research use only