Learn More
Medchemexpress LLC P-CAB agent 2 hydrochloride | 2209911-80-6 | C22H26ClFN2O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1532195MG
Encompass_Preferred
P-CAB agent 2 hydrochloride is a potent and orally active potassium-competitive acid blocker and a gastric acid secretion inhibitor. It inhibits H+/K+-ATPase activity with an IC50 value of <100 nM and the hERG potassium channel with an IC50 value of 18.69 μM. It demonstrates no acute toxicity and inhibits histamine-induced gastric acid secretion in SD rats.
  • Potent and orally active
  • Inhibits H+/K+-ATPase activity
  • Inhibits hERG potassium channel
  • Shows no acute toxicity
  • Inhibits histamine-induced gastric acid secretion
  • Shipped at room temperature in continental US
  • Powder stable for up to 3 years at -20°C
  • Solution stable for up to 6 months at -80°C
Catalog No. 50-003-72575