Learn More
Medchemexpress LLC P-CAB agent 2 hydrochloride | 2209911-80-6 | MFCD18878621 | C22H26ClFN2O4S | 25 MG
SDP
Catalog No. 5000375623
Encompass_Preferred
P-CAB agent 2 hydrochloride is a potent and orally active potassium-competitive acid blocker and a gastric acid secretion inhibitor. It inhibits H+/K+-ATPase activity with an IC50 value of <100 nM. It also inhibits the hERG potassium channel with an IC50 value of 18.69 μM. This compound shows no acute toxicity and inhibits histamine-induced gastric acid secretion.
  • Potent and orally active potassium-competitive acid blocker
  • Gastric acid secretion inhibitor
  • Inhibits H+/K+-ATPase activity with an IC50 value of <100 nM
  • Inhibits the hERG potassium channel with an IC50 value of 18.69 μM
  • Shows no acute toxicity
  • Inhibits histamine-induced gastric acid secretion
Catalog No. 50-003-75623 Supplier Medchemexpress LLC Supplier No. 25MGHY153219
P-CAB agent 2 hydrochloride is a potent and orally active potassium-competitive acid blocker and a gastric acid secretion inhibitor. It inhibits H+/K+-ATPase activity with an IC50 value of <100 nM. It also inhibits the hERG potassium channel with an IC50 value of 18.69 μM. This compound shows no acute toxicity and inhibits histamine-induced gastric acid secretion.
  • Potent and orally active potassium-competitive acid blocker
  • Gastric acid secretion inhibitor
  • Inhibits H+/K+-ATPase activity with an IC50 value of <100 nM
  • Inhibits the hERG potassium channel with an IC50 value of 18.69 μM
  • Shows no acute toxicity
  • Inhibits histamine-induced gastric acid secretion