Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human P-cadherin (CDH3) ELISA Kit

Catalog No. EHCDH3X10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHCDH3X10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EHCDH3X10 Supplier Invitrogen™ Supplier No. EHCDH3X10
Add to Cart
Add to Cart
Request a Quote

ELISA

The Human P-Cadherin (CDH3) ELISA quantitates Hu P-Cadherin in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu P-Cadherin. Principle of the method The Human P-Cadherin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

P-cadherin, also known as Cadherin-3, is a Type 1 cadherin protein that belongs to the cadherin superfamily. Type 1 cadherins are single-pass transmembrane proteins that have 5 extracellular cadherin repeats and an intracellular domain that binds p120-catenin and beta-catenin. Cadherins are calcium-dependent cell-cell adhesion glycoproteins responsible for a range of processes including development, wound healing, cell-cell signaling, cell growth and differentiation. P-cadherin is expressed in human fetal structures, and in adult tissues such as the basal layer of the epidermis, breast, prostate, ovary, cervix, hair follicle, and corneal endothelium. P-cadherin expression has also been reported on embryonic stem cells, and stem cells of the normal mammary gland, and hair follicle. Overexpression of P-cadherin has been associated with poor prognosis in breast, prostate, ovary, colon, and stomach carcinomas. Mutations in CDH3 are associated with congential hypotrichosis with juvenile macular dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P22223
Assay Range 20-18000 pg/mL
Assay Sensitivity 20 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Growth Factors & Receptors
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AI385538; CADH3; cadherin 15, M-cadherin (myotubule); cadherin 3; cadherin 3, type 1, P-cadherin (placental); cadherin-3; Cadp; calcium-dependent adhesion protein, placental; CDH3; CDHP; HJMD; PCAD; P-cadherin; Placental cadherin; Placental-cadherin; RPE-specific cadherinAI385538; CADH3; cadherin 15, M-cadherin (myotubule); cadherin 3; cadherin 3, type 1, P-cadherin (placental); cadherin-3; Cadp; calcium-dependent adhesion protein, placental; CDH3; CDHP; HJMD; PCAD; P-cadherin; Placental cadherin; Placental-cadherin; RPE-specific cadherin; CDHP; HJMD; PCAD
Gene ID (Entrez) 1001
Gene Symbol Cdh3
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Placental cadherin, P-cadherin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 3.3 μL, Serum 3.3 μL, Supernatant 100 μL
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target P-cadherin
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.