Learn More
Bioss P-Cadherin - N-terminal region
SDP
Catalog No. 5000065109
Encompass_Preferred
P-Cadherin (N-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB M596Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65109 Supplier Bioss Supplier No. BSM70369M
P-Cadherin (N-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB M596Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol