Learn More
Cayman Chemical p-chloro Phenylalanine, Purity: ≥98%, CAS: 7424-00-2, 25 g
SDP
Catalog No. 502935084
Encompass_Preferred
An inhibitor of tryptophan hydroxylase (Ki = 300 µM); a competitive inhibitor in vitro and an irreversible inhibitor in vivo; depletes serotonin to undetectable levels in rats after six days at 300 mg/kg; has commonly been used to deplete brain serotonin levels in animal studies of the serotonergic system
Catalog No. 50-293-5084 Supplier Cayman Chemical Supplier No. 2616825 G
An inhibitor of tryptophan hydroxylase (Ki = 300 µM); a competitive inhibitor in vitro and an irreversible inhibitor in vivo; depletes serotonin to undetectable levels in rats after six days at 300 mg/kg; has commonly been used to deplete brain serotonin levels in animal studies of the serotonergic system