Learn More
Bioworld p-Chlorophenoxyacetic Acid (CPA), 25 g
Catalog No. 50255040
Encompass_Preferred
p-Chlorophenoxyacetic acid (CPA)Synonym: 4-CPA Influence cell enlargement, root initiation and adventitious bud formation Appearance: off-white to tan crystals Melting Point: 157-159 °CSoluble in etanol and KOH Harmful if swallowed
Catalog No. 50-255-040 Supplier Bioworld Supplier No. 306310071
p-Chlorophenoxyacetic acid (CPA)Synonym: 4-CPA Influence cell enlargement, root initiation and adventitious bud formation Appearance: off-white to tan crystals Melting Point: 157-159 °CSoluble in etanol and KOH Harmful if swallowed