Learn More
Cayman Chemical p-Coumaric Acid, Purity: ≥98%, CAS: 501-98-4, 100 g
SDP
Catalog No. 502983519
Encompass_Preferred
A hydroxy derivative of cinnamic acid that can be found in a variety of edible plants that is reported to have antioxidant, anti-inflammatory, and antimicrobial activity
Catalog No. 50-298-3519 Supplier Cayman Chemical Supplier No. 20929100 G
A hydroxy derivative of cinnamic acid that can be found in a variety of edible plants that is reported to have antioxidant, anti-inflammatory, and antimicrobial activity