Learn More
Medchemexpress LLC HY-N0351 100mg Medchemexpress, p-Coumaric acid CAS:501-98-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0351100MG
Encompass_Preferred
Medchemexpress, HY-N0351 100mg p-Coumaric acid CAS:501-98-4 p-Coumaric acid is the abundant isomer of cinnamic acid which has antitumor and anti-mutagenic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3532