Learn More
Medchemexpress LLC p-Coumaric acid-13C3 | 1261170-80-2 | 167.14 | 1 MG
SDP
Catalog No. 5000389336
Encompass_Preferred
p-Coumaric acid-13C3 is the 13C-labeled version of p-Coumaric acid. p-Coumaric acid is a prevalent isomer of cinnamic acid, known for its antitumor and anti-mutagenic properties. This compound is intended for research use only and is not sold to patients. Stable heavy isotopes, including carbon, are incorporated into drug molecules primarily as tracers for quantification during drug development. Deuteration, in particular, has gained attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89336 Supplier Medchemexpress LLC Supplier No. HYN0351S1MG
p-Coumaric acid-13C3 is the 13C-labeled version of p-Coumaric acid. p-Coumaric acid is a prevalent isomer of cinnamic acid, known for its antitumor and anti-mutagenic properties. This compound is intended for research use only and is not sold to patients. Stable heavy isotopes, including carbon, are incorporated into drug molecules primarily as tracers for quantification during drug development. Deuteration, in particular, has gained attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.