Learn More
Medchemexpress LLC p-Coumaric acid-13C3 | 1261170-80-2 | 167.14 | 100 UG
SDP
Catalog No. 5000390069
Encompass_Preferred
p-Coumaric acid-13C3 is a 13C-labeled version of p-Coumaric acid, an abundant isomer of cinnamic acid known for its antitumor and anti-mutagenic activities. Stable heavy isotopes have been incorporated into this compound, primarily for use as tracers in drug development.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-90069 Supplier Medchemexpress LLC Supplier No. HYN0351S100UG
p-Coumaric acid-13C3 is a 13C-labeled version of p-Coumaric acid, an abundant isomer of cinnamic acid known for its antitumor and anti-mutagenic activities. Stable heavy isotopes have been incorporated into this compound, primarily for use as tracers in drug development.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.