Learn More
Medchemexpress LLC p-Coumaric acid-13C3 | 1261170-80-2 | 167.14 | 500 UG
SDP
Catalog No. 5000390080
Encompass_Preferred
p-Coumaric acid-13C3 is the 13C-labeled p-Coumaric acid, an abundant isomer of cinnamic acid known for its antitumor and anti-mutagenic activities.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Purity of 99.12%.
  • Appears as a solid, white to off-white.
Catalog No. 50-003-90080 Supplier Medchemexpress LLC Supplier No. HYN0351S500UG
p-Coumaric acid-13C3 is the 13C-labeled p-Coumaric acid, an abundant isomer of cinnamic acid known for its antitumor and anti-mutagenic activities.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Purity of 99.12%.
  • Appears as a solid, white to off-white.