Learn More
Selleck Chemical LLC p-Coumaric Acid S4759-1g
SDP
Catalog No. 503111969
Encompass_Preferred
p-Coumaric acid (4-Hydroxycinnamic acid P-Hydroxycinnamic acid 4-Coumaric acid Trans-p-Coumaric acid para-Coumaric Acid) is a hydroxy derivative of cinnamic acid found in a variety of edible plants and is reported to have antioxidant anti-inflammatory and antimicrobial activity
Catalog No. 50-311-1969 Supplier Selleck Chemical LLC Supplier No. S47591G
p-Coumaric acid (4-Hydroxycinnamic acid P-Hydroxycinnamic acid 4-Coumaric acid Trans-p-Coumaric acid para-Coumaric Acid) is a hydroxy derivative of cinnamic acid found in a variety of edible plants and is reported to have antioxidant anti-inflammatory and antimicrobial activity