Learn More
Medchemexpress LLC p-Coumaric acid | 501-98-4 | 164.16 | 1 ML
SDP
Catalog No. 5000390173
Encompass_Preferred
p-Coumaric acid (trans-4-Hydroxycinnamic acid) is an isomer of cinnamic acid with oral activity.
  • Inhibits cell proliferation and promotes apoptosis.
  • Exhibits antibacterial, anti-inflammatory, antioxidant, and anti-tumor activities.
  • Can significantly inhibit the proliferation of human and mouse melanoma cells.
  • Shows antibacterial activity against gram-negative and gram-positive bacteria, with MIC values for Escherichia coli, Streptococcus dysenteriae, and Salmonella typhimurium of 80, 10, and 20 μg/ml respectively.
  • Has a protective effect on colon pretumor induced by 1,2 dimethylhydrazine (DMH) in rats.
  • Alleviates nephrotoxicity induced by Doxorubicin by inhibiting oxidative stress, inflammation, and apoptosis.
Catalog No. 50-003-90173 Supplier Medchemexpress LLC Supplier No. HYN035110MM/1ML
p-Coumaric acid (trans-4-Hydroxycinnamic acid) is an isomer of cinnamic acid with oral activity.
  • Inhibits cell proliferation and promotes apoptosis.
  • Exhibits antibacterial, anti-inflammatory, antioxidant, and anti-tumor activities.
  • Can significantly inhibit the proliferation of human and mouse melanoma cells.
  • Shows antibacterial activity against gram-negative and gram-positive bacteria, with MIC values for Escherichia coli, Streptococcus dysenteriae, and Salmonella typhimurium of 80, 10, and 20 μg/ml respectively.
  • Has a protective effect on colon pretumor induced by 1,2 dimethylhydrazine (DMH) in rats.
  • Alleviates nephrotoxicity induced by Doxorubicin by inhibiting oxidative stress, inflammation, and apoptosis.