Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Coumaric Acid, 98+%, (Crystalline), MP Biomedicals

Catalog No. ICN10257610 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10257610 10 g
ICN10257601 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10257610 Supplier MP Biomedicals, Inc Supplier No. 0210257610
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Predominantly trans-isomer

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7400-08-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
InChI Key NGSWKAQJJWESNS-ZZXKWVIFSA-N
Synonym p-coumaric acid, 4-hydroxycinnamic acid, p-hydroxycinnamic acid, 4-coumaric acid, trans-4-hydroxycinnamic acid, trans-p-coumaric acid, p-cumaric acid, 3-4-hydroxyphenyl acrylic acid, para-coumaric acid, hydroxycinnamic acid
PubChem CID 637542
ChEBI CHEBI:32374
IUPAC Name (E)-3-(4-hydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=CC=C1C=CC(=O)O)O

Specifications

Assay Percent Range 98+%
Quantity 10 g
Formula Weight 164.2
Physical Form Crystalline
Chemical Name or Material p-Coumaric Acid
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.