Learn More
Medchemexpress LLC p-Coumaric acid ethyl ester | 7362-39-2 | MFCD00210460 | 50 MG
SDP
Catalog No. 5000421278
Encompass_Preferred
p-Coumaric Acid Ethyl Ester is a non-competitive, reversible inhibitor of tyrosinase. It quenches the intrinsic fluorescence of the enzyme and changes the binding affinity of L-tyrosine by inducing conformational changes in the catalytic domain of tyrosinase, without binding to the copper ion of the enzyme. This compound is utilized in the development of medicines, cosmetics, and for fruit preservation.
  • Non-competitive, reversible inhibitor of tyrosinase
  • Quenches the intrinsic fluorescence of the enzyme
  • Induces conformational changes in the catalytic domain of tyrosinase without binding to copper ion
  • Exhibits anti-platelet activity in rabbit platelet-rich plasma
  • Utilized in medicine and cosmetic development
  • Used for fruit preservation
Catalog No. 50-004-21278 Supplier Medchemexpress LLC Supplier No. HYN310350MG
p-Coumaric Acid Ethyl Ester is a non-competitive, reversible inhibitor of tyrosinase. It quenches the intrinsic fluorescence of the enzyme and changes the binding affinity of L-tyrosine by inducing conformational changes in the catalytic domain of tyrosinase, without binding to the copper ion of the enzyme. This compound is utilized in the development of medicines, cosmetics, and for fruit preservation.
  • Non-competitive, reversible inhibitor of tyrosinase
  • Quenches the intrinsic fluorescence of the enzyme
  • Induces conformational changes in the catalytic domain of tyrosinase without binding to copper ion
  • Exhibits anti-platelet activity in rabbit platelet-rich plasma
  • Utilized in medicine and cosmetic development
  • Used for fruit preservation