Learn More
Medchemexpress LLC p-Coumaric acid (Standard) | 501-98-4 | 164.16 | 50 MG
SDP
Catalog No. 5000390171
Encompass_Preferred
p-Coumaric acid (Standard) is the analytical standard of p-Coumaric acid, intended for research and analytical applications. It is an isomer of cinnamic acid with oral activity. p-Coumaric acid inhibits cell proliferation and promotes apoptosis. It also has antibacterial, anti-inflammatory, antioxidant, and anti-tumor activities.
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Exhibits antibacterial activity
  • Exhibits anti-inflammatory activity
  • Exhibits antioxidant activity
  • Exhibits anti-tumor activity
Catalog No. 50-003-90171 Supplier Medchemexpress LLC Supplier No. HYN0351R50MG
p-Coumaric acid (Standard) is the analytical standard of p-Coumaric acid, intended for research and analytical applications. It is an isomer of cinnamic acid with oral activity. p-Coumaric acid inhibits cell proliferation and promotes apoptosis. It also has antibacterial, anti-inflammatory, antioxidant, and anti-tumor activities.
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Exhibits antibacterial activity
  • Exhibits anti-inflammatory activity
  • Exhibits antioxidant activity
  • Exhibits anti-tumor activity