Learn More
Medchemexpress LLC p-Coumaric acid (trans-4-hydroxycinnamic acid) | 501-98-4 | 164.16 | 10 G
SDP
Catalog No. 5000389744
Encompass_Preferred
p-Coumaric acid, an isomer of cinnamic acid, demonstrates diverse biological activities including antibacterial, anti-inflammatory, antioxidant, and anti-tumor effects. It has been shown to inhibit cell proliferation and promote apoptosis, making it a subject of interest in various research applications.
  • Inhibits proliferation of A375 and B16 cells.
  • Promotes cell apoptosis in human and mouse melanoma cells.
  • Exhibits antibacterial activity against gram-negative and gram-positive bacteria.
  • Reduces polyp incidence and precancerous lesions in animal models.
  • Alleviates doxorubicin-induced nephrotoxicity by inhibiting oxidative stress, inflammation, and apoptosis.
  • Modulates cell cycle progression and protein expression in A375 and B16 cells.
Catalog No. 50-003-89744 Supplier Medchemexpress LLC Supplier No. HYN035110G
p-Coumaric acid, an isomer of cinnamic acid, demonstrates diverse biological activities including antibacterial, anti-inflammatory, antioxidant, and anti-tumor effects. It has been shown to inhibit cell proliferation and promote apoptosis, making it a subject of interest in various research applications.
  • Inhibits proliferation of A375 and B16 cells.
  • Promotes cell apoptosis in human and mouse melanoma cells.
  • Exhibits antibacterial activity against gram-negative and gram-positive bacteria.
  • Reduces polyp incidence and precancerous lesions in animal models.
  • Alleviates doxorubicin-induced nephrotoxicity by inhibiting oxidative stress, inflammation, and apoptosis.
  • Modulates cell cycle progression and protein expression in A375 and B16 cells.