Learn More
Medchemexpress LLC p-Coumaric acid (trans-4-Hydroxycinnamic acid) | 501-98-4 | 164.16 | 500 MG
SDP
Catalog No. 5000389759
Encompass_Preferred
p-Coumaric acid (trans-4-Hydroxycinnamic acid) is an isomer of cinnamic acid with oral activity that exhibits several biological activities:
  • Inhibits cell proliferation
  • Promotes apoptosis
  • Has antibacterial activity
  • Has anti-inflammatory activity
  • Has antioxidant activity
  • Has anti-tumor activities
Catalog No. 50-003-89759 Supplier Medchemexpress LLC Supplier No. HYN0351500MG
p-Coumaric acid (trans-4-Hydroxycinnamic acid) is an isomer of cinnamic acid with oral activity that exhibits several biological activities:
  • Inhibits cell proliferation
  • Promotes apoptosis
  • Has antibacterial activity
  • Has anti-inflammatory activity
  • Has antioxidant activity
  • Has anti-tumor activities