Learn More
Medchemexpress LLC P-cresol glucuronide | 17680-99-8 | MFCD08277028 | 284.26 g/mol | C13H16O7 | 1 MG
SDP
Catalog No. 5000281616
Encompass_Preferred
p-Cresol glucuronide is the glucuronide conjugate of p-cresol and is described as a protein-bound uremic toxin associated with chronic kidney disease. It is supplied as a research reagent and reference standard for studies of drug metabolism, microbiome-derived metabolites, and kidney disease pathophysiology.
  • Useful as a reference standard for analytical and metabolic studies.
  • Relevant for research into uremic toxins and chronic kidney disease.
  • Suitable for use in in vitro and analytical assays requiring small-quantity standards.
  • Typically accompanied by product documentation such as datasheets and handling notes.
  • Store under recommended conditions to preserve chemical stability.
Catalog No. 50-002-81616 Supplier Medchemexpress LLC Supplier No. HYW4162501MG
p-Cresol glucuronide is the glucuronide conjugate of p-cresol and is described as a protein-bound uremic toxin associated with chronic kidney disease. It is supplied as a research reagent and reference standard for studies of drug metabolism, microbiome-derived metabolites, and kidney disease pathophysiology.
  • Useful as a reference standard for analytical and metabolic studies.
  • Relevant for research into uremic toxins and chronic kidney disease.
  • Suitable for use in in vitro and analytical assays requiring small-quantity standards.
  • Typically accompanied by product documentation such as datasheets and handling notes.
  • Store under recommended conditions to preserve chemical stability.