Learn More
Cayman Chemical p-Cresol GlucuronIde 1mg
SDP
Catalog No. 502910501
Encompass_Preferred
A metabolite of p-cresol; formed from p-cresol primarily by UGT1A6, but also by UGT1A9, in human liver and kidney microsomes; accumulates in feces, blood, and urine of patients with CKD
Catalog No. 50-291-0501 Supplier Cayman Chemical Supplier No. 351131 MG
A metabolite of p-cresol; formed from p-cresol primarily by UGT1A6, but also by UGT1A9, in human liver and kidney microsomes; accumulates in feces, blood, and urine of patients with CKD