Learn More
Cayman Chemical p-Cresol sulfatepotasIM250mg
SDP
Catalog No. 502990165
Encompass_Preferred
A sulfate conjugate of p-cresol; increases migration and proliferation of isolated rat aortic VSMCs at 500 µM; increases aortic atherosclerotic plaques lesion area in nephrectomized ApoE-/- mice fed a high-fat diet when administered at 100 mg/kg per day via the drinking water; serum levels are increased in patients with advanced stage chronic kidney disease and positively associated with vascular calcification
Catalog No. 50-299-0165 Supplier Cayman Chemical Supplier No. 29504250 MG
A sulfate conjugate of p-cresol; increases migration and proliferation of isolated rat aortic VSMCs at 500 µM; increases aortic atherosclerotic plaques lesion area in nephrectomized ApoE-/- mice fed a high-fat diet when administered at 100 mg/kg per day via the drinking water; serum levels are increased in patients with advanced stage chronic kidney disease and positively associated with vascular calcification