Learn More
Apexbio Technology LLC p-Cresyl sulfate, 3233-58-7, MFCD27964764, 10 mg
SDP
Catalog No. 501151154
Encompass_Preferred
MF: C7H8O4S, MW: 188.20, Solubility: 30.1mg/mL in DMSO, 50mg/mL in H2O. The p-Cresol is a protein-bound uremic retention solute.
Catalog No. 50-115-1154 Supplier Apexbio Technology LLC Supplier No. A889510
MF: C7H8O4S, MW: 188.20, Solubility: 30.1mg/mL in DMSO, 50mg/mL in H2O. The p-Cresol is a protein-bound uremic retention solute.