Learn More
Medchemexpress LLC p-Cresyl sulfate | 3233-58-7 | 100.0% | 188.20 | 1 ML
SDP
Catalog No. 5000345713
Encompass_Preferred
p-Cresyl sulfate (p-Tolyl sulfate) is a uremic toxin known to cause renal damage and dysfunction. It exhibits antiproliferation activity and influences the expression of HIF-1α, VHL, and HIF-2α. This compound also induces epithelial-mesenchymal transition (EMT) and activates the JNK and p38 MAPK signaling pathways.
  • Shows antiproliferation activity.
  • Increases protein expression of HIF-1α and VHL.
  • Decreases protein expression of HIF-2α.
  • Induces epithelial-mesenchymal transition (EMT).
  • Activates JNK and p38 MAPK signaling pathways.
Catalog No. 50-003-45713 Supplier Medchemexpress LLC Supplier No. HY11143110MG
p-Cresyl sulfate (p-Tolyl sulfate) is a uremic toxin known to cause renal damage and dysfunction. It exhibits antiproliferation activity and influences the expression of HIF-1α, VHL, and HIF-2α. This compound also induces epithelial-mesenchymal transition (EMT) and activates the JNK and p38 MAPK signaling pathways.
  • Shows antiproliferation activity.
  • Increases protein expression of HIF-1α and VHL.
  • Decreases protein expression of HIF-2α.
  • Induces epithelial-mesenchymal transition (EMT).
  • Activates JNK and p38 MAPK signaling pathways.