Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC p-Cresyl sulfate | 3233-58-7 | MFCD27964764 | 99.9% | 188.20 g/mol | C7H8O4S | 5 MG
SDP

Catalog No. 5000263502
Encompass_Preferred

p-Cresyl sulfate is a small-molecule uremic toxin supplied as a solid or ready-to-use solution for research use. It is provided as a high-purity analytical standard suitable for biochemical assays, cell-based studies, and analytical method development.

  • High purity (99.87%).
  • Physical form: white to off-white solid; also available as DMSO solution.
  • Molecular formula C7H8O4S; molecular weight 188.20 g/mol.
  • Soluble in DMSO (250 mg/mL) and water (≥100 mg/mL).
  • Store at 4°C protected from light; in solution store at -80°C (6 months) or -20°C (1 month).
  • Suitable for biochemical and cell-based assays, and analytical method development.

Catalog No. 50-002-63502 Supplier Medchemexpress LLC Supplier No. HY1114315MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

p-Cresyl sulfate is a small-molecule uremic toxin supplied as a solid or ready-to-use solution for research use. It is provided as a high-purity analytical standard suitable for biochemical assays, cell-based studies, and analytical method development.

  • High purity (99.87%).
  • Physical form: white to off-white solid; also available as DMSO solution.
  • Molecular formula C7H8O4S; molecular weight 188.20 g/mol.
  • Soluble in DMSO (250 mg/mL) and water (≥100 mg/mL).
  • Store at 4°C protected from light; in solution store at -80°C (6 months) or -20°C (1 month).
  • Suitable for biochemical and cell-based assays, and analytical method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.