Learn More
TARGETMOL CHEMICALS INC P-CRESYL SULFATE 5MG
SDP
Catalog No. 502577200
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. p-Cresyl Sulfate is a major uremic toxin found in the blood of patients with chronic kidney disease (CKD) derived from the metabolites of tyrosine and phenylalanine in the liver. purity: 97%
Catalog No. 50-257-7200 Supplier TARGETMOL CHEMICALS INC Supplier No. T219775MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. p-Cresyl Sulfate is a major uremic toxin found in the blood of patients with chronic kidney disease (CKD) derived from the metabolites of tyrosine and phenylalanine in the liver. purity: 97%