Learn More
Apexbio Technology LLC p-Cresyl sulfate potassium 91978-69-7 50mg
SDP
Catalog No. 5000094563
Encompass_Preferred
p-Cresyl sulfate potassium (CAS 91978-69-7) is the potassium salt form of p-cresyl sulfate a metabolite that accumulates as a protein-bound uremic toxin in patients with chronic kidney disease Biologically it exerts proinflammatory effects by activating cellular signaling pathways including c-Jun N-terminal kinase (JNK) and p38 mitogen-activated protein kinase (MAPK) which can enhance inflammatory responses Research indicates that p-cresyl sulfate potassium also contributes to cardiovascular risk by adversely impacting vascular and cardiac function This compound is utilized in experimental studies investigating the molecular mechanisms of uremic toxicity inflammation and cardiorenal syndromes
Catalog No. 50-000-94563 Supplier Apexbio Technology LLC Supplier No. A992550
p-Cresyl sulfate potassium (CAS 91978-69-7) is the potassium salt form of p-cresyl sulfate a metabolite that accumulates as a protein-bound uremic toxin in patients with chronic kidney disease Biologically it exerts proinflammatory effects by activating cellular signaling pathways including c-Jun N-terminal kinase (JNK) and p38 mitogen-activated protein kinase (MAPK) which can enhance inflammatory responses Research indicates that p-cresyl sulfate potassium also contributes to cardiovascular risk by adversely impacting vascular and cardiac function This compound is utilized in experimental studies investigating the molecular mechanisms of uremic toxicity inflammation and cardiorenal syndromes