Learn More
Medchemexpress LLC p-Cresyl sulfate potassium | 91978-69-7 | 99.9% | 226.29 | 50 MG
SDP
Catalog No. 5000345534
Encompass_Preferred
p-Cresyl sulfate potassium (Standard) is an analytical standard used in research and analytical applications. It is a uremic toxin linked to renal damage and dysfunction. This compound exhibits antiproliferation activity, influences protein expression (HIF-1α, VHL, HIF-2α), and activates JNK and p38 MAPK signaling pathways. It is suitable for experiments involving HPLC, GC, and MS.
  • Analytical standard for research.
  • Uremic toxin causing renal damage and dysfunction.
  • Exhibits antiproliferation activity.
  • Modulates protein expression (HIF-1α, VHL, HIF-2α).
  • Induces epithelial-mesenchymal transition (EMT).
  • Activates JNK and p38 MAPK signaling pathways.
  • Applicable in HPLC, GC, and MS research.
Catalog No. 50-003-45534 Supplier Medchemexpress LLC Supplier No. HY111431AR50MG
p-Cresyl sulfate potassium (Standard) is an analytical standard used in research and analytical applications. It is a uremic toxin linked to renal damage and dysfunction. This compound exhibits antiproliferation activity, influences protein expression (HIF-1α, VHL, HIF-2α), and activates JNK and p38 MAPK signaling pathways. It is suitable for experiments involving HPLC, GC, and MS.
  • Analytical standard for research.
  • Uremic toxin causing renal damage and dysfunction.
  • Exhibits antiproliferation activity.
  • Modulates protein expression (HIF-1α, VHL, HIF-2α).
  • Induces epithelial-mesenchymal transition (EMT).
  • Activates JNK and p38 MAPK signaling pathways.
  • Applicable in HPLC, GC, and MS research.