Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich p-Cymene

Catalog No. C1214521L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1214521L 1 L
C1214524L 4 L
C12145225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C1214521L Supplier Sigma Aldrich Supplier No. C1214521L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

p-Cymene is an alkyl aromatic hydrocarbon mainly used to study oxidation reactions under various conditions to synthesize highly valuable intermediates and building-blocks such as cymene hydroperoxide, 4-methyl acetophenone, 4-isopropyl benzyl alcohol, 4-isopropyl benzaldehyde and 4-isopropyl benzoic acid etc.

Specifications

CAS 99-87-6
Density 0.86 g/mL (at 25°C (literature))
Boiling Point 176°C to 178°C (lit.)
Molecular Formula C10H14
Refractive Index n20/D 1.490 (literature)
Linear Formula CH3C6H4CH(CH3)2
MDL Number MFCD00008893
Quantity 1 L
Synonym 1-Isopropyl-4-methylbenzene; 4-Isopropyltoluene
Molecular Weight (g/mol) 134.22
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.