Learn More
Medchemexpress LLC HY-Y0015 500mg Medchemexpress, p-Dimethylaminobenzaldehyde CAS:100-10-7 Purity:>98%
Supplier: Medchemexpress LLC HYY0015500MG
Medchemexpress, HY-Y0015 500mg p-Dimethylaminobenzaldehyde CAS:100-10-7 p-Dimethylaminobenzaldehyde (4-Dimethylaminobenzaldehyde) is an organic compound containing amine and aldehyde moieties which is used in Ehrlich's reagent and Kovac's reagent to test for indoles. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.