Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Dimethylaminobenzaldehyde, 5% in 1-Propanol, Ricca Chemical
SDP

Catalog No. R25545001C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R25545001C 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R25545001C Supplier Ricca Chemical Company Supplier No. R25545001C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71-23-8 , 100-10-7
Molecular Formula C3H8O
Molecular Weight (g/mol) Mixture
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

CAS Min % 92.0, 5.76
CAS Max % 96.0, 5.99
Packaging Amber Glass Bottle
Color Colorless to Yellow
Quantity 1 L
Physical Form Liquid
Percent Purity 5% W/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 5% in 1-Propanol
Chemical Name or Material p-Dimethylaminobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.