Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Dithiane-2,5-diol, 96%

Catalog No. AC179930010
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC179930010 1 kg Plastic bottle
AC179932500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC179930010 Supplier Thermo Scientific Chemicals Supplier No. 179930010
Only null left
Add to Cart

CAS: 40018-26-6 | C4H8O2S2 | 152.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40018-26-6
Molecular Formula C4H8O2S2
Molecular Weight (g/mol) 152.23
InChI Key YUIOPHXTILULQC-UHFFFAOYSA-N
Synonym p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anme
PubChem CID 98330
IUPAC Name 1,4-dithiane-2,5-diol
SMILES C1C(SCC(S1)O)O

Specifications

Melting Point 130.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Plastic bottle
Quantity 1 kg
Solubility Information Solubility in water: 3.9g/L (20°C).
Formula Weight 152.23
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material p-Dithiane-2,5-diol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic if swallowed.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 254-751-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.