Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Dithiane-2,5-diol, 96%, Acros Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Plastic bottle
40018-26-6
C4H8O2S2
152.23
YUIOPHXTILULQC-UHFFFAOYSA-N
p-dithiane-2,5-diol,2,5-dihydroxy-1,4-dithiane,mercaptoacetaldehyde dimer,4-dithiane-2,5-diol,1,4 dithiane-2,5-diol,2,5-dihydroxy-1,4-dithian,p-dthane-2,5-dol,pubchem20480,1,4dithiane-2,5diol,acmc-1anme
98330
1,4-dithiane-2,5-diol
C1C(SCC(S1)O)O
40018-26-6
C4H8O2S2
152.23
YUIOPHXTILULQC-UHFFFAOYSA-N
p-dithiane-2,5-diol,2,5-dihydroxy-1,4-dithiane,mercaptoacetaldehyde dimer,4-dithiane-2,5-diol,1,4 dithiane-2,5-diol,2,5-dihydroxy-1,4-dithian,p-dthane-2,5-dol,pubchem20480,1,4dithiane-2,5diol,acmc-1anme
98330
1,4-dithiane-2,5-diol
C1C(SCC(S1)O)O
Melting Point 130.0°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 250g
Color White to Yellow
Assay Percent Range 95% min. (GC)
Solubility Information Solubility in water: 3.9g/L (20°C).
Formula Weight 152.23
Physical Form Crystalline Powder
Percent Purity 96%
Chemical Name or Material p-Dithiane-2,5-diol
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic if swallowed.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

EINECSNumber : 254-751-6